Thermo Scientific Chemicals

Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041675.JU
1 mol, Each
Thermo Scientific Chemicals

Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 041675.JU
1 mol, Each
Quantity
Catalog number: 041675.JU
also known as 041675-JU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
925-90-6
IUPAC Name
bromo(ethyl)magnesium
Molecular Formula
C2H5BrMg
InChI Key
TWTWFMUQSOFTRN-UHFFFAOYSA-M
SMILES
CC[Mg]Br
Form
Liquid
Appearance (Color)
Clear to slightly hazy
Assay from Supplier's CofA
2.85-3.30M

Description

Ethylmagnesium bromide is used as a reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text