Thermo Scientific Chemicals

Lithium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041832.18
50 g, Each
Thermo Scientific Chemicals

Lithium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041832.18
50 g, Each
Quantity
Catalog number: 041832.18
also known as 041832-18
Price (USD)
Price: 298.00
Online price: 253.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12057-24-8
IUPAC Name
dilithium(1+) hydroxide
Molecular Formula
HLi2O
InChI Key
XUCJHNOBJLKZNU-UHFFFAOYSA-M
SMILES
[Li+].[Li+].[OH-]
Total Metal Impurities
0.5% max.

Description

Ceramics and special glass formulations, carbon dioxide absorbent.Lithium oxide is used as a flux in ceramic glazes, co-dopant with yttria in the zirconia ceramic top coat and as the cathode in the lithium ion batteries, which is used in power electronic devices like mobile phones, laptop computers and battery-powered cars. It is also used to prepare lithium hydroxide and lithium metal by electrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Air and Moisture Sensitive; Readily absorbs carbon dioxide and water from air
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text