Thermo Scientific Chemicals

Tetraammineplatinum(II) hydroxide solution, Pt 8-11% w/w (cont. Pt), Thermo Scientific Chemicals

Catalog number: 042918.NH
1 g, Each
Thermo Scientific Chemicals

Tetraammineplatinum(II) hydroxide solution, Pt 8-11% w/w (cont. Pt), Thermo Scientific Chemicals

Catalog number: 042918.NH
1 g, Each
Quantity
Catalog number: 042918.NH
also known as 042918-NH
Price (USD)
Price: 303.00
Online price: 257.65
Your price:
Quantity
-

Chemical Identifiers

CAS
38201-97-7
IUPAC Name
platinum(2+) tetraamine dihydroxide
Molecular Formula
H14N4O2Pt
InChI Key
VSKCDODNDPOZKS-UHFFFAOYSA-L
SMILES
N.N.N.N.[OH-].[OH-].[Pt++]
Concentration (weight/weight)
Pt 8-11%

Description

Tetraammineplatinum(II) hydroxide solution acts as a precursor for a platinum catalyst. It acts as noble metal catalyst which is primarily useful for homogeneous catalysis applications. It is used as a catalyst in hydrogenation, carbonylation, hydroformylation, chiral catalysis reactions and ruthenium dyes. Furthermore, it is used for C-X coupling reaction like Heck, Suzuki and Stille reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text