Thermo Scientific Chemicals

cis-4-Octene, 97%, Thermo Scientific Chemicals

Catalog number: 043156.18
50 g, Each
Thermo Scientific Chemicals

cis-4-Octene, 97%, Thermo Scientific Chemicals

Catalog number: 043156.18
50 g, Each
Quantity
Catalog number: 043156.18
also known as 043156-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7642-15-1
Appearance (Color)
Colorless
Assay (GC)
≥96%
GC/MS Analysis
Conforms
Form
Liquid

Description

cis-4-octene is used as a chain transfer agent in polymerization reactions, and as a depolymerizing agent in epoxy-functionalized oligomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text