Thermo Scientific Chemicals

Mesoporphyrin IX dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 043320.MD
250 mg, Each
Thermo Scientific Chemicals

Mesoporphyrin IX dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 043320.MD
250 mg, Each
Quantity
Catalog number: 043320.MD
also known as 043320-MD
Price (USD)
Price: 420.00
Online price: 357.65
Your price:
Quantity
-

Chemical Identifiers

CAS
68938-72-7
Appearance (Color)
Dark red
Assay from Supplier's CofA
≥96.0% (HPLC)
Form
Powder or crystalline powder
Proton NMR
Conforms to structure

Description

Mesoporphyrin IX dihydrochloride is used reactant in constructing efficient dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text