Thermo Scientific Chemicals

Tellurium(IV) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043439.18
50 g, Each
Thermo Scientific Chemicals

Tellurium(IV) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043439.18
50 g, Each
Quantity
Catalog number: 043439.18
also known as 043439-18
Price (USD)
Price: 624.00
Online price: 540.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10031-27-3
Residual water
< 100 ppm
Form
Powder
Total Metal Impurities
0.001% max.
Appearance (Color)
Orange

Description

Tellurium tetrabromide it acts as a chemical and organic intermediate. It is also acts a conductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text