Thermo Scientific Chemicals

Bis(methylcyclopentadienyl)niobium dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 044393.03
1 g, Each
Thermo Scientific Chemicals

Bis(methylcyclopentadienyl)niobium dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 044393.03
1 g, Each
Quantity
Catalog number: 044393.03
also known as 044393-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
61374-51-4
Molecular Formula
C12H12Cl2Nb
InChI Key
SZUFSQYBUPXGSX-UHFFFAOYSA-N
SMILES
Cl.Cl.C[c-]1cccc1.C[C-]12[C-]3[C-]4[C-]5[C-]1[Nb]2345
Molecular Weight (g/mol)
320.04
Appearance (Color)
Brown to dark brown
Form
Powder
Purity
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text