Thermo Scientific Chemicals

Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 044434.22
100 g, Each
Thermo Scientific Chemicals

Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 044434.22
100 g, Each
Quantity
Catalog number: 044434.22
also known as 044434-22
Price (USD)
Price: 458.00
Online price: 388.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3396-11-0
IUPAC Name
caesium(1+) acetate
Molecular Formula
C2H3CsO2
InChI Key
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES
[Cs+].CC([O-])=O
Total Metal Impurities
0.002% max.

Description

Cesium acetate is used in organic synthesis especially in the Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text