Thermo Scientific Chemicals
Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals
Catalog number: 044692.06
5 g, Each
Thermo Scientific Chemicals
Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals
Catalog number: 044692.06
5 g, Each
Quantity
Catalog number: 044692.06
also known as 044692-06
Price (USD)
Price: 36.80
Online price: 31.65
Your price:
Quantity
-
Description
Ethyl hydrogen malonate is used as a reactant for the preparation of tetramic acids through Dieckmann ring closure and organocatalytic decarboxylative Doebner-Knoevenagel reactions. It is involved in the acylation reactions and Knoevenagel condensation with aldehydes. It is also used in the preparation of gamma-lactones from olefins by intermolecular carbolactonization in presence of Mn(III) acetate as a catalyst.