Thermo Scientific Chemicals

Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 044711.14
25 g, Each
Thermo Scientific Chemicals

Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 044711.14
25 g, Each
Quantity
Catalog number: 044711.14
also known as 044711-14
Price (USD)
Price: 101.00
Online price: 85.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3279-54-7
IUPAC Name
disodium phenyl phosphate
Molecular Formula
C6H5Na2O4P
InChI Key
TYJOJLOWRIQYQM-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5%
Form
Crystalline powder
Water Content (Karl Fischer Titration)
11 - 17%
Assay (HPLC)
≥97.5%

Description

Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text