Thermo Scientific Chemicals

Poly(3-octylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Catalog number: 044777.03
1 g, Each
Thermo Scientific Chemicals

Poly(3-octylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Catalog number: 044777.03
1 g, Each
Quantity
Catalog number: 044777.03
also known as 044777-03
Price (USD)

Chemical Identifiers

CAS
104934-51-2
Molecular Formula
(C12H18S)n
SMILES
CCCCCCCCC1=C(-*)SC(-*)=C1
MDL Number
MFCD00241412
Assay (unspecified)
Mw 30,000 to 50,000
lambda-max (UV-Vis)
Absorption ( in THF) 440 to 450 nm
Trace Metals (by AAS)
Nickel < 100 ppm
Form
Fibers
Assay (unspecified)
Polydispersity: 2 to 4

Description

It is employed Conducting polymer, rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials. It is also used in organic field-effect transistors and in polymer-based solar cells and p-type polymer semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text