Thermo Scientific Chemicals

Polydimethylsiloxane (silicone) emulsion, Thermo Scientific Chemicals

Catalog number: 045088.22
100 g, Each
Thermo Scientific Chemicals

Polydimethylsiloxane (silicone) emulsion, Thermo Scientific Chemicals

Catalog number: 045088.22
100 g, Each
Quantity
Catalog number: 045088.22
also known as 045088-22
Price (USD)
Price: 24.40
Online price: 18.65
Your price:
Quantity
-
Form
Liquid
Appearance (Color)
White
Assay (unspecified)
54.0-64.0% solids content

Description

Polydimethylsiloxane (silicone) emulsion is the most widely used silicon-based organic polymer and is particularly known for its unusual rheological (or flow) properties. Further, it is used in contact lenses, medical devices to elastomers and in shampoos (as dimethicone makes hair shiny and slippery). It finds application as an antifoaming agent in food, caulking, lubricants, kinetic sand and heat-resistant tiles. In addition to this, it serves as a critical ingredient in water-repelling coatings such as Rain-X.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text