Thermo Scientific Chemicals

Rhenium(VII) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 045538.04
2 g, Each
Thermo Scientific Chemicals

Rhenium(VII) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 045538.04
2 g, Each
Quantity
Catalog number: 045538.04
also known as 045538-04
Price (USD)
Price: 182.00
Online price: 154.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-68-7
IUPAC Name
trioxo[(trioxorhenio)oxy]rhenium
Molecular Formula
O7Re2
InChI Key
NBGOSNNAAHDRLK-UHFFFAOYSA-N
SMILES
O=[Re](=O)(=O)O[Re](=O)(=O)=O
Appearance (Color)
Yellow to black
Form
Crystalline
Total Metal Impurities
≤0.005% (50 ppm)

Description

Rhenium(VII) oxide is a precursor to organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is also used as a catalyst for hemiacetal substitution and as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text