Thermo Scientific Chemicals

Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals

Catalog number: 045896.A4
2.5 kg, Each
Thermo Scientific Chemicals

Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals

Catalog number: 045896.A4
2.5 kg, Each
Quantity
Catalog number: 045896.A4
Price (USD)
Price: 808.00
Online price: 686.65
Your price:
Quantity
-

Chemical Identifiers

CAS
68083-14-7
IUPAC Name
3,3-dimethyl-5,5-diphenyl-2,4-dioxa-3,5-disilahexane
Molecular Formula
C16H22O2Si2
InChI Key
ARWRSWALIGRKQA-UHFFFAOYSA-N
SMILES
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1
Form
clear, viscous liquid
Refractive Index
1.500 to 1.510 ( at 25 deg C)
Appearance (Color)
Colorless
Assay (unspecified)
% volatile ( 24 hours, @ 150 deg C):

Description

Silicone oil is used in high temperature open systems, closed systems and circulating, closed loop and heat transfer baths. It is used as a high temperature fluid for laboratory research apparatus and instruments. It is also used as a calibration media in temperature controlled baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High temperature open system, closed system, circulating, closed loop, and heat transfer baths
  • High temperature fluids for laboratory research apparatus and instruments
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text