Thermo Scientific Chemicals

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0), Thermo Scientific Chemicals

Catalog number: 045913.MD
250 mg, Each
Thermo Scientific Chemicals

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0), Thermo Scientific Chemicals

Catalog number: 045913.MD
250 mg, Each
Quantity
Catalog number: 045913.MD
also known as 045913-MD
Price (USD)

Chemical Identifiers

CAS
565441-56-7
IUPAC Name
Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0)
Molecular Formula
C96H94Fe2P2Pd
SMILES
[Fe].[Fe].[Pd].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)
1527.87
Appearance (Color)
Pink
Assay (unspecified)
5.70-6.70% Hydrogen
Assay (unspecified)
74.50-76.50% Carbon
Form
Solid

Description

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0) is used to prepare palladium(0) catalysts for highly selective cross-coupling reactions especially carbohalogenation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text