Product Image
Thermo Scientific™

Dionex™ SolEx™ HRP Columns, 2.1 x 20mm, 12-14μm

Designed for fast, easy preconcentration of contaminants from water samples prior to HPLC analysis.
Have Questions?
Catalog NumberDiameter (Metric) Outer
0755902.1 mm
Catalog number 075590
Price (USD)
412.95
Each
Diameter (Metric) Outer:
2.1 mm
Price (USD)
412.95
Each

Thermo Scientific™ Dionex™ SolEx™ on-line solid-phase extraction (SPE) Cartridges are designed for fast, easy preconcentration of contaminants from water samples prior to HPLC analysis.

On-line SolEx HRP Cartridges

High capacity with good recoveries Wide pH compatibility (0 to 14) Packed with a divinylbenzene polymer with a hydrophilic bonded layer Resin is used in the reversed-phase mode and demonstrates excellent retention properties on both polar and hydrophobic analytes Cartridge fits in a holder for easy exchange when necessary

Recommended for: Pharmaceutical and pesticides in water supplies, or endocrine disruptors in seawater can be concentrated with this cartridge, prior to HPLC separation.
Specifications
Diameter (Metric) Outer2.1 mm
For Use With (Application)On-line HPLC
Length (Metric)20 mm
MaterialHydrophilic Divinylbenzene Polymer
QuantityEach
Product LineSolEx
pH0 to 14
Unit SizeEach