Thermo Scientific Chemicals

Thallium(III) nitrate trihydrate, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 087999.22
100 g, Each
Thermo Scientific Chemicals

Thallium(III) nitrate trihydrate, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 087999.22
100 g, Each
Quantity
Catalog number: 087999.22
also known as 087999-22
Price (USD)
Price: 1,181.00
Online price: 1,022.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13453-38-8
Total Metal Impurities
0.5% max.

Description

Thallium(III) nitrate trihydrate is a reagent used for the rapid oxidation of styrene derivatives to carbonyl compounds, for the regioselective ring expansion of cyclic aralkyl ketones via oxidative rearrangement, for the selective hydrolysis of thioacetals to carbonyl compounds and for a general synthetic method to 19-norsteroids. Mediates ring contraction of cyclic ketones to a series of trans-fused 10-methyl-2-decalones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text