Thermo Scientific Chemicals

Sodium tetraethylborate, 98%, Thermo Scientific Chemicals

Catalog number: 089137.06
5 g, Each
Thermo Scientific Chemicals

Sodium tetraethylborate, 98%, Thermo Scientific Chemicals

Catalog number: 089137.06
5 g, Each
Quantity
Catalog number: 089137.06
also known as 089137-06
Price (USD)
Price: 662.00
Online price: 562.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15523-24-7
IUPAC Name
sodium tetraethylboranuide
Molecular Formula
C8H20BNa
InChI Key
SZSBMTRYJRHYNI-UHFFFAOYSA-N
SMILES
[Na+].CC[B-](CC)(CC)CC
Appearance (Color)
white
Form
Powder
Elemental Analysis
>97.5%

Description

Sodium tetraethylborate is used as a derivatizing agent for the detection of organotin, lead and mercury compounds by using gas chromatography coupled with various sensitive and selective detection methods, such as atomic absorption spectrometry (AAS), atomic emission spectrometry (AES), pulsed-flame photometric detection (PFPD), and mass spectrometry (MS). It is also used as an alkylating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text