Thermo Scientific Chemicals

Ferrocene, 98%, Thermo Scientific Chemicals

Catalog number: 119145000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ferrocene, 98%, Thermo Scientific Chemicals

Catalog number: 119145000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 119145000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
102-54-5
IUPAC Name
Ferrocene
Molecular Formula
C10H10Fe
InChI Key
DFRHTHSZMBROSH-UHFFFAOYSA-N
SMILES
[Fe].c1cccc1.c1cccc1
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Color)
Orange-brown
Appearance
May become lumpy on storage
Melting point
173°C to 176°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text