Thermo Scientific Chemicals

Guanidine Carbonate, 99+%, Thermo Scientific Chemicals

Catalog number: 120220051
5 kg, Each
Thermo Scientific Chemicals

Guanidine Carbonate, 99+%, Thermo Scientific Chemicals

Catalog number: 120220051
5 kg, Each
Quantity

Chemical Identifiers

CAS
593-85-1
IUPAC Name
carbonic acid; bis(guanidine)
Molecular Formula
C3H12N6O3
InChI Key
STIAPHVBRDNOAJ-UHFFFAOYSA-N
SMILES
NC(N)=N.NC(N)=N.OC(O)=O
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 %
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text