Thermo Scientific Chemicals

2-Iodoaniline, 98%, Thermo Scientific Chemicals

Catalog number: 122301000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Iodoaniline, 98%, Thermo Scientific Chemicals

Catalog number: 122301000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122301000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-43-0
Appearance (Color)
White to yellow to beige to brown
Appearance (Form)
Powder or crystals or needles
Infrared spectrum
Conforms
Melting point
53°C to 58°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text