Thermo Scientific Chemicals

Methyl 5-bromovalerate, 97%, Thermo Scientific Chemicals

Catalog number: 126460250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 5-bromovalerate, 97%, Thermo Scientific Chemicals

Catalog number: 126460250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 126460250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5454-83-1
IUPAC Name
methyl 5-bromopentanoate
Molecular Formula
C6H11BrO2
InChI Key
RAVVJKCSZXAIQP-UHFFFAOYSA-N
SMILES
COC(=O)CCCCBr
Appearance (Form)
Liquid
Refractive index
1.4620 to 1.4640 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Color)
Clear colorless to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text