Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 130135000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 130135000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130135000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5144-89-8
IUPAC Name
1,10-phenanthroline hydrate
Molecular Formula
C12H10N2O
InChI Key
PPQJCISYYXZCAE-UHFFFAOYSA-N
SMILES
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Appearance (Color)
White to cream
Appearance (Form)
Crystalline powder or needles
Infrared spectrum
Conforms
Titration with HClO4
99.0 to 101.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text