Thermo Scientific Chemicals

4-Bromo-2-fluoroaniline, 99%, Thermo Scientific Chemicals

Catalog number: 134180250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromo-2-fluoroaniline, 99%, Thermo Scientific Chemicals

Catalog number: 134180250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
367-24-8
IUPAC Name
4-bromo-2-fluoroaniline
Molecular Formula
C6H5BrFN
InChI Key
GZRMNMGWNKSANY-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=C1F
Appearance (Form)
chunks
Appearance (Color)
Beige to grey to light brown
Melting point
38°C to 42°C
Appearance (Form)
Crystalline powder or crystals or mass and/or
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text