Thermo Scientific Chemicals

alpha,alpha,alpha-Trifluorotoluene, 99+%, Thermo Scientific Chemicals

Catalog number: 139775000
500 mL, Plastic bottle, Each
Thermo Scientific Chemicals

alpha,alpha,alpha-Trifluorotoluene, 99+%, Thermo Scientific Chemicals

Catalog number: 139775000
500 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 139775000
Price (USD)
Quantity
-

Chemical Identifiers

CAS
98-08-8
IUPAC Name
(trifluoromethyl)benzene
Molecular Formula
C7H5F3
InChI Key
GETTZEONDQJALK-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %
Refractive index
1.4130 to 1.4150 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text