Thermo Scientific Chemicals

N-Acetylsulfanilyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 150725000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Acetylsulfanilyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 150725000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 150725000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-60-8
IUPAC Name
4-acetamidobenzene-1-sulfonyl chloride
Molecular Formula
C8H8ClNO3S
InChI Key
GRDXCFKBQWDAJH-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O
Appearance (Form)
Crystalline or granular powder or crystals
Infrared spectrum
Conforms
Melting point
144°C to 148°C (Dec)
Titration Argentometric
>=98.5 %
Appearance (Color)
White to cream to light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text