Thermo Scientific Chemicals

Ethoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 159860250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 159860250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159860250
Price (USD)
Price: 203.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
627-03-2
IUPAC Name
2-ethoxyacetic acid
Molecular Formula
C4H8O3
InChI Key
YZGQDNOIGFBYKF-UHFFFAOYSA-N
SMILES
CCOCC(O)=O
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4170 to 1.4200 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text