Thermo Scientific Chemicals

1,3,5,7-Cyclooctatetraene, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 160570100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

1,3,5,7-Cyclooctatetraene, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 160570100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 160570100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-20-9
IUPAC Name
(1Z,3Z,5Z,7Z)-cycloocta-1,3,5,7-tetraene
Molecular Formula
C8H8
InChI Key
KDUIUFJBNGTBMD-DLMDZQPMSA-N
SMILES
C1=C/C=C\C=C/C=C\1
Appearance (Color)
Turbid yellow to yellow-brown
Appearance (Form)
Liquid
Refractive index
1.5365 to 1.5385 (20°C, 589 nm)
Stabilizer
0.1 % hydroquinone
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text