Thermo Scientific Chemicals

Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194350500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194350500
50 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
16962-40-6
IUPAC Name
titanium(4+) diammonium hexafluoride
Molecular Formula
F6H8N2Ti
InChI Key
NMGYKLMMQCTUGI-UHFFFAOYSA-J
SMILES
[NH4+].[NH4+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
Appearance (Form)
Crystalline powder or crystals and/or chunks
Total trace metal impurities
=<0.01 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text