Thermo Scientific Chemicals

hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals

Catalog number: 195370250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals

Catalog number: 195370250
25 g, Glass Bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
26023-84-7
IUPAC Name
dihydrogen hexachloroplatinumtetrakis(ylium)
Molecular Formula
Cl6H2Pt
InChI Key
ZKOQTCCVSRSECD-UHFFFAOYSA-J
SMILES
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
Total trace metal impurities
=<0.1 %
Assay
37.5 to 40.5 % (Pt)
Appearance (Form)
Powder and/or chunks
Appearance (Color)
Orange to orange-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text