Thermo Scientific Chemicals

Ruthenium(IV) oxide hydrate, > 54% Ru, Thermo Scientific Chemicals

Catalog number: 197430250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Ruthenium(IV) oxide hydrate, > 54% Ru, Thermo Scientific Chemicals

Catalog number: 197430250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
32740-79-7
IUPAC Name
dioxoruthenium
Molecular Formula
O2Ru
InChI Key
WOCIAKWEIIZHES-UHFFFAOYSA-N
SMILES
O=[Ru]=O
Assay
>=54 % (Ru)
Appearance (Color)
Black
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text