Thermo Scientific™

Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers

Catalog number: 2118CV
Thermo Scientific™

Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers

Catalog number: 2118CV
Keep your oxygen scavenger analyzer ready to use with Thermo Scientific™ Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers. Choose from options including: oxygen scavenger consumables kit; reagent-less oxygen scavenger consumables kit; fluid connector block assembly with thumb screw and injection port; flow cell assembly with electrode holder, mounting bracket and O-rings; and mixing coil-tubing spool assembly.
 
Catalog Number
2118CV
Unit Size
Each
Description
Calibration valve assembly
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionCalibration valve assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeValve Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Oxygen scavenger consumables kit
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionOxygen scavenger consumables kit
TypeConsumables Kit
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Includes6 x 1L iodide reagent and 6 x 6 ft. diffusion tubing assemblies with O-rings, reagent bottle adapter, 1 pint of 100ppm hydrazine standard, iodide electrode, reference electrode, 5 x 2 oz. reference electrode filling solution, O-ring kit, restriction tube
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reagent-less oxygen scavenger consumables kit
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionReagent-less oxygen scavenger consumables kit
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeConsumables Kit
Includes6 x 1L empty bottles for iodide reagent, 6 x 6 ft. diffusion tubing assemblies with O-rings (181860), reagent bottle adapter (2118RBA), 1 pint of 100 ppm hydrazine standard (151810), iodide electrode (100022), reference electrode (100056), 5 x 2 oz. reference electrode filling solution (181073), O-ring kit (2118OK), restriction tube assembly (2100RT), 2 x 60 micron inlet filters and gasket (181170)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Fluid connector block assembly with thumb screw and injection port
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionFluid connector block assembly with thumb screw and injection port
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeBlock Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Flow cell assembly with electrode holder, mounting bracket and O-rings
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionFlow cell assembly with electrode holder, mounting bracket and O-rings
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeFlow-cell Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Includes2118XP Flow Cell Assembly with Electrode Holder, Mounting Bracket and O-rings
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mixing coil-tubing spool assembly
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionMixing coil-tubing spool assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeSpool Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Micro-valve assembly
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionMicro-valve assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeValve Assembly
For Use With (Application)Calibration standard injection port
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reagent bottle adapter assembly with diffusion tube assembly
Price (USD)
Price: 381.00
Your price:
Availability
-
Full specifications
Quantity
DescriptionReagent bottle adapter assembly with diffusion tube assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeAdapter Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
2118XP Electrode Kit - includes iodide electrode, reference electrode and 2 oz. reference electrode filling solution
Price (USD)
Availability
-
Full specifications
Quantity
-
Description2118XP Electrode Kit - includes iodide electrode, reference electrode and 2 oz. reference electrode filling solution
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeElectrode Kit
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
IncludesIodide electrode, reference electrode and 2 oz. reference electrode filling solution
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Electronics faceplate
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionElectronics faceplate
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeElectronics Faceplate
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Showing 10 of 14Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
2118CVFull specifications
EachCalibration valve assemblyRequest A Quote--
DescriptionCalibration valve assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeValve Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
211850XPFull specifications
EachOxygen scavenger consumables kitRequest A Quote--
DescriptionOxygen scavenger consumables kit
TypeConsumables Kit
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Includes6 x 1L iodide reagent and 6 x 6 ft. diffusion tubing assemblies with O-rings, reagent bottle adapter, 1 pint of 100ppm hydrazine standard, iodide electrode, reference electrode, 5 x 2 oz. reference electrode filling solution, O-ring kit, restriction tube
Unit SizeEach
211850XPFFull specifications
EachReagent-less oxygen scavenger consumables kitRequest A Quote--
DescriptionReagent-less oxygen scavenger consumables kit
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeConsumables Kit
Includes6 x 1L empty bottles for iodide reagent, 6 x 6 ft. diffusion tubing assemblies with O-rings (181860), reagent bottle adapter (2118RBA), 1 pint of 100 ppm hydrazine standard (151810), iodide electrode (100022), reference electrode (100056), 5 x 2 oz. reference electrode filling solution (181073), O-ring kit (2118OK), restriction tube assembly (2100RT), 2 x 60 micron inlet filters and gasket (181170)
Unit SizeEach
2118CBFull specifications
EachFluid connector block assembly with thumb screw and injection portRequest A Quote--
DescriptionFluid connector block assembly with thumb screw and injection port
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeBlock Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
2118FCFull specifications
EachFlow cell assembly with electrode holder, mounting bracket and O-ringsRequest A Quote--
DescriptionFlow cell assembly with electrode holder, mounting bracket and O-rings
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeFlow-cell Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Includes2118XP Flow Cell Assembly with Electrode Holder, Mounting Bracket and O-rings
Unit SizeEach
2118MCFull specifications
EachMixing coil-tubing spool assemblyRequest A Quote--
DescriptionMixing coil-tubing spool assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeSpool Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
2118MVFull specifications
EachMicro-valve assemblyRequest A Quote--
DescriptionMicro-valve assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeValve Assembly
For Use With (Application)Calibration standard injection port
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
2118RBAFull specifications
EachReagent bottle adapter assembly with diffusion tube assembly
Price: 381.00
Your price:
-
DescriptionReagent bottle adapter assembly with diffusion tube assembly
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeAdapter Assembly
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
2118XPEKFull specifications
Each2118XP Electrode Kit - includes iodide electrode, reference electrode and 2 oz. reference electrode filling solutionRequest A Quote--
Description2118XP Electrode Kit - includes iodide electrode, reference electrode and 2 oz. reference electrode filling solution
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeElectrode Kit
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
IncludesIodide electrode, reference electrode and 2 oz. reference electrode filling solution
Unit SizeEach
2118XPEPFull specifications
EachElectronics faceplateRequest A Quote--
DescriptionElectronics faceplate
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeElectronics Faceplate
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
Unit SizeEach
Showing 10 of 14Show more
Keep your oxygen scavenger analyzer ready to use with Thermo Scientific™ Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers. Choose from options including: oxygen scavenger consumables kit; reagent-less oxygen scavenger consumables kit; fluid connector block assembly with thumb screw and injection port; flow cell assembly with electrode holder, mounting bracket and O-rings; and mixing coil-tubing spool assembly.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text