Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Dispense strong acids/alkalis, reagents and solvents with Thermo Scientific™ Nalgene™ Drop-Dispensing Bottle made with Teflon™ FEP with ETFE Dropping Closure and Cap, featuring excellent chemical resistance for dispensing virtually any chemical. Tethered spout cap snaps securely into place and won't pop off or leak when the closed bottle is squeezed. Usable over a wide temperature range -105°C to +150°C. Leakproof† and autoclavable.
Closure Material | ETFE |
Closure Type | Cap |
Through the Closure Dispensing | Yes |
Product Type | Wash Bottle |
Capacity | 1 oz, 30mL |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | Translucent |
Description | Teflon FEP Drop-Dispensing Bottle with Dropping Closure and Cap |
Material | FEP |
No. per Pack | 1 |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Pack of 1 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2414-0030PK also known as 2414-0030 | Pack of 1 | 30 mL | 151.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Ordering Information:
One per package; case of four.