Thermo Scientific Chemicals

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog number: 243100250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog number: 243100250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 243100250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82911-69-1
IUPAC Name
2,5-dioxopyrrolidin-1-yl (9H-fluoren-9-yl)methyl carbonate
Molecular Formula
C19H15NO5
InChI Key
WMSUFWLPZLCIHP-UHFFFAOYSA-N
SMILES
O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)ON1C(=O)CCC1=O
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
HPLC
>=97.5 %
Melting point
145°C to 153°C
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text