Thermo Scientific Chemicals

(S)-N-FMOC-Octylglycine, 95%, 98% ee

Catalog number: 361490010
Each
Thermo Scientific Chemicals

(S)-N-FMOC-Octylglycine, 95%, 98% ee

Catalog number: 361490010
Each
Catalog number: 361490010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
193885-59-5
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)decanoic acid
Molecular Formula
C25H31NO4
InChI Key
LSMLSLHVRMSYPT-UHFFFAOYNA-N
SMILES
CCCCCCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Enantiomeric excess
>=97.5 %
HPLC
>=94 %
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text