The dC(bz) ABI LV40™ columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.
• Proprietary design and manufacturing methods ensure consistency and high quality • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free • Hydrophobic polystryrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis • The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length
Save Time and Money Compared to conventional columns, the ABI LV40™ columns reduce cycle time by about 10% and the cost of synthesis by about 25%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column Type
Oligonucleotide Synthesis Column
Format
Column
Sample Type
Oligos
For Use With (Equipment)
3900 DNA Synthesizer
For Use With (Application)
Oligo Synthesis
Matrix
Polystyrene
Pore Size
1000 Å
Prep Scale
40 nmol
Product Line
LV40™
Product Type
Oligonucleotide Synthesis Column
Shipping Condition
Room Temperature
Target
Nucleic Acids
Contents & Storage
Columns may be stored in a dry environment at room temperature.