Applied Biosystems™

ABI LV40™ dGdmf Pack

Catalog number: 4316344
Applied Biosystems™

ABI LV40™ dGdmf Pack

Catalog number: 4316344
Catalog Number
4316344
Unit Size
60 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
431634460 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Contains 60 dG(dmf) ABI LV40™ columns.

The dG(dmf) ABI LV40™ columns are fully assembled, low-cost columns that produce high-quality oligonucleotides. ABI LV40™ columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.

• Proprietary design and manufacturing methods ensure consistency and high quality
• Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
• Hydrophobic polystryrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis
• The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length

Save Time and Money
Compared to conventional columns, the ABI LV40™ columns reduce cycle time by about 10% and the cost of synthesis by about 25%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type
Oligonucleotide Synthesis Column
Format
Column
Sample Type
Oligos
For Use With (Equipment)
3900 DNA Synthesizer
For Use With (Application)
Oligo Synthesis
Matrix
Polystyrene
Pore Size
1000 Å
Prep Scale
40 nmol
Product Line
LV40™
Product Type
Oligonucleotide Synthesis Column
Shipping Condition
Room Temperature
Target
Nucleic Acids

Contents & Storage

Columns may be stored in a dry environment at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text