See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
Home
Shop All Products
Clinical Analyzers and Instruments
DNA Sequencing Systems
GeneMapper™
ID-X
Software v1.1, Full Installation
Applied Biosystems™
GeneMapper™
ID-X
Software v1.1, Full Installation
Catalog number:
4404049
Related applications:
Forensics
Technical Support
Customer Service
Applied Biosystems™
GeneMapper™
ID-X
Software v1.1, Full Installation
Catalog number:
4404049
Related applications:
Forensics
Technical Support
Customer Service
Catalog Number
4404049
Unit Size
1 license
Price (USD)
Price:
60,000.00
Your price:
Sign In
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
4404049
1 license
Price:
60,000.00
Your price:
Sign In
-
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This kit includes one full installation of GeneMapper™ ID-X Software v1.1 including database. Documentation included.
Monitor
Recommended Configuration (Client Installation):
• 1024 x 768 pixel resolution.
• 12-inch color monitor (smaller monitor lessens ability to view a full screen of data).
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line
GeneMapper™
Software Type
Forensic STR Fragment Analysis
For Use With (Equipment)
310 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730 DNA Analyzer, 3130 Genetic Analyzer
Operating System
Windows 2000 Pro (Service Pack 4), Windows XP (Service Pack 2)
Product Type
Software V1.1
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate