Thermo Scientific Chemicals

Acetamiprid

Catalog number: 463930010
1 g, Each
Thermo Scientific Chemicals

Acetamiprid

Catalog number: 463930010
1 g, Each
Quantity

Chemical Identifiers

CAS
135410-20-7
IUPAC Name
N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
Molecular Formula
C10H11ClN4
InChI Key
WCXDHFDTOYPNIE-UHFFFAOYSA-N
SMILES
CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N
Appearance (Form)
Powder
Melting point
95°C to 102°C
HPLC
>=95.0 %
Appearance (Color)
White to light yellow
Water
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text