Search Thermo Fisher Scientific
Use Thermo Scientific™ Cimarec™ Extension Cables with Cimarec Magnetic Stirrers.
Description | 8-Pin Extension Cable |
For Use With (Equipment) | Telemodul 40 M and 80 M |
Length (Metric) | 3 m |
Type | Extension Cable |
Unit Size | Each |
Description | 4-Pin Extension Cable |
For Use With (Equipment) | Telemodul 20 C, 40 C and 10 M |
Length (Metric) | 3 m |
Type | Extension Cable |
Unit Size | Each |
Description | 6-Pin Extension Cable |
For Use With (Equipment) | Biomodul 40 B |
Length (Metric) | 3 m |
Type | Extension Cable |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
50088102 | Each | 8-Pin Extension Cable | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
50088016 | Each | 4-Pin Extension Cable | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
50088021 | Each | 6-Pin Extension Cable | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|