MSC-Advantage™ Class II Biological Safety Cabinets
Thermo Scientific™

MSC-Advantage™ Class II Biological Safety Cabinets

Conserve energy with these cabinets, which combine smart design and extraordinary value with best-in-class energy efficiency, reliability and usability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (English) InteriorWidth (Metric) Interior
510254114 ft.1.2 m
510254136 ft.1.8 m
510282252.95 ft.0.9 m
510282264.92 ft.1.5 m
Catalog number 51025411
Price (HKD)
-
Width (English) Interior:
4 ft.
Width (Metric) Interior:
1.2 m

Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets (BSCs) combine safety and extraordinary value with energy efficiency, reliability and usability. With proprietary airflow design and energy-efficient DC motors, the MSC-Advantage BSCs use up to 68% less energy than cabinets with traditional AC motors. The cabinets provide both personal and product protection from biological hazards and contamination while affording users with a comfortable and ergonomic workspace. Cabinets are certified to EN 12469 safety standard.

The MSC-Advantage biological safety is available in 4 sizes and can be connected to an external exhaust to provide protection for trace amounts of volatile toxic chemicals that are sometimes used in the cabinet.

  • Available in 90 cm, 120 cm, 150 cm and 180 cm

Containment

  • Thermo Scientific™ SmartFlow™ technology automates airflow balancing and compensation
  • Digital Airflow Verification (DAVe) alerts user to compromised product and operator protection
  • H14 HEPA filters remove airborne contaminants
  • SmartFlow™ indicator displays performance status

Comfort

  • Low noise level enhances user focus
  • 10° sloped front for exceptional comfort and ergonomics
  • Spacious work area with comfortable armrests
  • Increased light intensity for a brighter work environment

Convenience 

  • SmartClean front window design to improve cleaning and loading of large equipment
  • Intuitive control panel displays safety and performance data
  • Thermo Scientific™ SmartPort™ design organises tubing and cables
  • Programmable UV light extends bulb life and saves energy (UV light is optional)

Certifications/Compliance

  • Fully compliant with the EN 12469 safety standard as independently tested and certified by TÜV Nord

Ordering Information:

Please order separately: Stand options, programmable UV light, one-piece work tray, ergonomic stool, free-standing floor shelf. Not available for sale in North America (equivalent model: 1300 Series A2 Biological Safety Cabinets).

Specifications
Depth (Metric) Exterior80 cm
Energy Consumption (Operating Set Point)200 W
Energy Consumption (Reduced Flow Mode)<40 W
Exhaust Air FilterHEPA H 14 EN 1822, 99.995% at 0.3μm Particle
Height (Metric) Exterior152 cm
PortsTUV-approved
Product TypeBiosafety Cabinet
Width (English) Exterior51.1 in.
Width (English) Interior4 ft.
Width (Metric) Exterior130 cm
Width (Metric) Interior1.2 m
Certifications/ComplianceEN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m)
Noise Level<59 dBA
Receptacles1 Double, Right Side
Voltage230 V
Unit SizeEach