Cytomat™ 2 C-LiN Automated Incubator
Cytomat™ 2 C-LiN Automated Incubator
Cytomat™ 2 C-LiN Automated Incubator
Cytomat™ 2 C-LiN Automated Incubator
Thermo Scientific™

Cytomat™ 2 C-LiN Automated Incubator

Temperature Range: Ambient +10°C to 50°C; Relative Humidity: <95%
Have Questions?
Catalog NumberElectrical RequirementsTemperature Range (Metric)
51032950100-230 V; 50/60 HzAmbient +10° to 50°C
Catalog number 51032950
Price (USD)
-
Electrical Requirements:
100-230 V; 50/60 Hz
Temperature Range (Metric):
Ambient +10° to 50°C

Ensure a safe, reproducible process, unsurpassed temperature uniformity and stability with the next-generation mid-range Thermo Scientific™ Cytomat™ 2 C-LiN series automated incubator.

This incubator is available in various temperature ranges and gate positions for hand-off to the lab automation robot. The device can be seamlessly integrated with a mean access time of 12 sec. As the first-generation mid-capacity automated incubator with a fully automated thermal decontamination and optional controlled humidity, the Cytomat 2 C-LiN series is designed to significantly decrease the risk of contamination as a new standard.

The Cytomat 2 C-LIN automated incubator provides an ideal safe environment for storage and incubation of assays in a temperature range of ambient +10° to 50°C, with high humidity up to 95% r.H. stable CO2 conditions and automated decontamination routine ContraCon without harmful gases.

Available Options

  • Integrated barcode reader enables sample tracking, barcode transmission, and inventory management.
  • P3-Plate detection in stacker helps to eliminate labware handling errors.
  • HydraSmart Technology for precise humidity control without heat extraction and external water tank to avoid contamination.
  • Right speed setting for the right application with individual speed setting per plate and adjustable incremental speed settings (Slow/Medium/Fast).
  • Flexible positioning inside your lab with 6 possible gate positions including underbench in a single body.
  • Solid Copper inner chamber for natural contamination prevention.

Key Features

  • Appropriate Temperature Distribution
  • Decontamination Routine
  • Fast and Reliable
  • Controlled Humidity
  • External Water Tank
Specifications
Access Time12 sec.
Certifications/ComplianceCE Marked
Depth (English)20.1 in.
Depth (Metric)511 mm
Electrical Requirements100-230 V; 50/60 Hz
FeaturesIncubation and Storage Unit; Humidity up to 95% r.H. CO2 range of 0-20 Vol. %; Includes decontamination routine-ContraCon
For Use With (Application)Incubation and storage for cell culture applications ambient +10°C to 50°C
Height (English)35.6 in.
Height (Metric)905 mm
Holds56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well
ModelCytomat 2 C-LiN Series Automated Incubator
TypeAutomated Incubator
Weight (Metric)80 kg
Width (English)22.5 in.
Width (Metric)572 mm
Chamber MaterialCopper
Relative Humidity<95%
Temperature Range (Metric)Ambient +10° to 50°C
Voltage100/230 V
Unit SizeEach