Search Thermo Fisher Scientific
Choose Thermo Scientific™ Dionex™ Flow Selectors for Thermo Scientific™ UltiMate™ NCS-3x00RS Nano/Capillary Pumps that are precalibrated to support given flow rate ranges for the UltiMate 3000 RSLCnano LC System.
Description | Flow Selector for NCS-3500 RS and NCP-3200 RS, for Micro LC with flow range 10-50 μL/min |
Max. Flow Rate | 10 μL/min. |
Type | Flow Selector |
For Use With (Equipment) | NCS-3500 RS and NCP-3200 RS, Micro LC |
Min. Flow Rate | 50 μL/min. |
Unit Size | Each |
Description | Flow Selector for NCS-3500 RS and NCP-3200 RS, for Nano LC with flow range 50-1000 nL/min |
Max. Flow Rate | 1000 nL/min. |
Type | Flow Selector |
For Use With (Equipment) | NCS-3500 RS and NCP-3200 RS, Nano LC |
Min. Flow Rate | 50 nL/min. |
Unit Size | Each |
Description | Flow Selector for NCS-3500 RS and NCP-3200 RS, for Capillary LC with flow range 0.5-10 μL/min |
Max. Flow Rate | 10 μL/min. |
Type | Flow Selector |
For Use With (Equipment) | NCS-3500 RS and NCP-3200 RS, Capillary LC |
Min. Flow Rate | 0.50 μL/min. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
6041.0014 | Each | NCS-3500 RS and NCP-3200 RS, Micro LC | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6041.0002 | Each | NCS-3500 RS and NCP-3200 RS, Nano LC | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6041.0003 | Each | NCS-3500 RS and NCP-3200 RS, Capillary LC | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Flow Selectors (recommended [settable]):