Thermo Scientific Chemicals

Cadmium, Granular, Thermo Scientific Chemicals

Catalog number: 612135000
500 g, Each
Thermo Scientific Chemicals

Cadmium, Granular, Thermo Scientific Chemicals

Catalog number: 612135000
500 g, Each
Quantity
Catalog number: 612135000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-43-9
IUPAC Name
cadmium
Molecular Formula
Cd
InChI Key
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES
[Cd]
Identification
passes test
Appearance (Color)
Silver white
Appearance (Form)
Granules

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text