Search Thermo Fisher Scientific
Thermo Scientific™ FT-IR Polymer Analysis Kits provide a rapid, inexpensive way to monitor chemical and physical parameters of polymers critical to your process or product quality. Whether you need to verify the correct co-polymer blend ratio or quantify the amount of release agent, UV stabilizer, or other additives in your materials, there is a polymer analysis configuration that’s right for you. The Polymer Analysis Kits can help ensure your raw materials, intermediates or finished products comply with your quality standards.
For Use With (Equipment) | Nicolet Summit Spectrometer |
Type | Basic Combo Polymer Kit |
Description | Nicolet Summit Basic Combo Polymer Kit |
Includes | OMNIC Specta Material Characterization for Polymer Labs, Nicolet Summit Everest ZnSe ATR, Mini Film Maker Kit (222-276700) NOTE: The Mini Film Maker Kit is used to prepare polymer films for FTIR transmission analysis. FT-IR transmission analysis requires the iD1 transmission accessory, order (840-319200). |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS5 |
Type | Basic ATR Polymer Kit |
Description | Basic ATR Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS20/iS50 |
Type | Basic ATR Polymer Kit |
Description | Basic ATR Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS5 |
Type | Advanced ATR Polymer Kit |
Description | Advanced ATR Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS20/iS50 |
Type | Advanced ATR Polymer Kit |
Description | Advanced ATR Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS5 |
Type | Basic Combo Polymer Kit |
Description | Basic Combo Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS20/iS50 |
Type | Basic Combo Polymer Kit |
Description | Basic Combo Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS5 |
Type | Advanced Combo Polymer Kit |
Description | Advanced Combo Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet iS20/iS50 |
Type | Advanced Combo Polymer Kit |
Description | Advanced Combo Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
For Use With (Equipment) | Nicolet Summit/iS5/iS20/iS50 |
Type | Mini-Film Maker Polymer Kit |
Description | Mini-Film Maker Polymer Kit |
For Use With (Application) | Polymer Analysis |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | For Use With (Equipment) | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
840-320300 | Each | Nicolet Summit Basic Combo Polymer Kit | Nicolet Summit Spectrometer | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-226400 | Each | Basic ATR Polymer Kit | Nicolet iS5 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-226900 | Each | Basic ATR Polymer Kit | Nicolet iS20/iS50 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-226500 | Each | Advanced ATR Polymer Kit | Nicolet iS5 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-227000 | Each | Advanced ATR Polymer Kit | Nicolet iS20/iS50 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-226700 | Each | Basic Combo Polymer Kit | Nicolet iS5 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-227200 | Each | Basic Combo Polymer Kit | Nicolet iS20/iS50 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-226800 | Each | Advanced Combo Polymer Kit | Nicolet iS5 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-227300 | Each | Advanced Combo Polymer Kit | Nicolet iS20/iS50 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-226600 | Each | Mini-Film Maker Polymer Kit | Nicolet Summit/iS5/iS20/iS50 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Thermo Scientific FT-IR Polymer Analysis Kits include either a single-bounce ATR accessory or a Heated Mini-Film Maker plus our unique Thermo Scientific OMNIC Specta Material Characterization software with a large spectral database of polymers and general chemical compounds. ATR allows you to quickly analyze polymer samples directly for identification or verification of bulk properties such as blend ratios. The Mini-Film Maker can be used to prepare constant thickness films from most thermoplastics for rapid and accurate quantitative analysis of polymer constituents and additives. The kits contain all of the components necessary to analyze your materials in your Thermo Scientific FT-IR spectrometer.
The Polymer Analysis Kit components:
OMNIC™ Specta* Polymer Package
ATR Accessory
Mini-Film Maker