Applications | Fluoride Analysis |
Connector Types | BNC, waterproof |
Description | Fluoride Ion Selective Electrode and Reagent Kit |
Diameter (English) Body | 0.55 in. |
Diameter (Metric) Body | 14mm |
Diameter (Metric) Cap | 17mm |
Filling Solution Type | Optimum Results A fill sol PN 90061 |
Internal Reference | 9606BNWP |
Junction | Sure-Flow |
Length (English) | 4.75 in. |
Length (English) Cable | 39.40 in. |
Length (Metric) | 110mm |
Length (Metric) Cable | 1m |
Material | Epoxy |
Operating Range (Metric) | RANGES: 1 x 10E-6M to saturated |
Temperature (English) Operating | 32-180°F |
Item Description | Electrode, Standards, ISA |
Membrane | pellet |
Model | 9606BNWP |
Electrode | Fluoride ISE |
Temperature (Metric) Range | 0-80°C |
Product Type | Kit |
For Use With (Equipment) | ISE Meter |
Temperature (Metric) Operating | 25°C |
Type | Fluoride Ion Selective Electrode and Reagent Kit |
Unit Size | Each |
Applications | Ammonia Analysis |
Connector Types | BNC, waterproof |
Description | HP Ammonia Ion Selective Electrode and Reagent Kit |
Diameter (English) Body | 0.47 in. |
Diameter (Metric) Body | 12mm |
Diameter (Metric) Cap | 16mm |
Filling Solution Type | HP Ammonia ISE fill sol PN 951209 |
Internal Reference | 9512HPBNWP |
Junction | Sure-Flow |
Length (English) | 4.75 in. |
Length (English) Cable | 39.40 in. |
Length (Metric) | 120mm |
Length (Metric) Cable | 1m |
Material | Epoxy |
Operating Range (Metric) | Ranges: 1.0 to 5 x 10E-7M |
Temperature (English) Operating | 32-122°F |
Item Description | Electrode, Standards, ISA |
Membrane | Ammonia Gas Sensing PN 951214 |
Response Time | < 2 min at 1 ppm and up |
Model | 9512HPBNWP |
Electrode | HP Ammonia ISE |
Temperature (Metric) Range | 0-50°C |
Product Type | Kit |
For Use With (Equipment) | ISE Meter |
Temperature (Metric) Operating | 25°C |
Type | Ammonia Ion Selective Electrode and Reagent Kit |
Unit Size | Each |
Applications | Ammonia Analysis |
Connector Types | BNC, waterproof |
Description | STD Ammonia Ion Selective Electrode and Reagent Kit |
Diameter (English) Body | 0.47 in. |
Diameter (Metric) Body | 12mm |
Diameter (Metric) Cap | 16mm |
Filling Solution Type | Std Ammonia ISE fill sol PN 951202 |
Internal Reference | 9512BNWP |
Junction | Sure-Flow |
Length (English) | 4.75 in. |
Length (English) Cable | 39.40 in. |
Length (Metric) | 150mm |
Length (Metric) Cable | 1m |
Material | Epoxy |
Operating Range (Metric) | Ranges : 1.0 to 5 x 10E-7M |
Temperature (English) Operating | 32-122°F |
Item Description | Electrode, Standards, ISA |
Membrane | Ammonia Gas Sensing PN 951204 |
Response Time | 1-4 min. |
Model | 9512BNWP |
Electrode | Ammonia ISE |
Temperature (Metric) Range | 0-50°C |
Product Type | Kit |
For Use With (Equipment) | ISE Meter |
Temperature (Metric) Operating | 25°C |
Type | Standard Ammonia ISE and Reagent Kit |
Unit Size | Each |
Applications | Fluoride Analysis |
Connector Types | BNC, waterproof |
Description | Low Level Fluoride Ion Selective Electrode and Reagent Kit |
Diameter (English) Body | 0.55 in. |
Diameter (Metric) Body | 14mm |
Diameter (Metric) Cap | 17mm |
Filling Solution Type | Optimum Results A fill sol PN 90061 |
Internal Reference | 9609BNWP |
Junction | Sure-Flow |
Length (English) | 4.75 in. |
Length (English) Cable | 39.40 in. |
Length (Metric) | 110mm |
Length (Metric) Cable | 1m |
Material | Epoxy |
Operating Range (Metric) | RANGES: 1 x 10E-6M to saturated |
Temperature (English) Operating | 32-180°F |
Item Description | Electrode,Standards, ISA |
Membrane | pellet |
Model | 9609BNWP |
Electrode | Fluoride ISE |
Temperature (Metric) Range | 0-80°C |
Product Type | Kit |
For Use With (Equipment) | ISE Meter |
Temperature (Metric) Operating | 25°C |
Type | Combination ISE and Reagent Kit |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Applications | Electrode | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
9609BNSLN | Each | Fluoride Ion Selective Electrode and Reagent Kit | Fluoride Analysis | Fluoride ISE | Price: 972.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9512HPBNSLN | Each | HP Ammonia Ion Selective Electrode and Reagent Kit | Ammonia Analysis | HP Ammonia ISE | Price: 993.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9512BNSLN | Each | STD Ammonia Ion Selective Electrode and Reagent Kit | Ammonia Analysis | Ammonia ISE | Price: 956.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
9609BNLSLN | Each | Low Level Fluoride Ion Selective Electrode and Reagent Kit | Fluoride Analysis | Fluoride ISE | Price: 1,031.00 Your Price: | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|