Thermo Scientific Chemicals

1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: A10154.30
250 g, Each
Thermo Scientific Chemicals

1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: A10154.30
250 g, Each
Quantity
Catalog number: A10154.30
also known as A10154-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-31-8
IUPAC Name
2-(piperazin-1-yl)ethan-1-amine
Molecular Formula
C6H15N3
InChI Key
IMUDHTPIFIBORV-UHFFFAOYSA-N
SMILES
NCCN1CCNCC1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4980-1.5020 @ 20?C

Description

1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text