Thermo Scientific Chemicals

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Catalog number: A10280.AP
500 mL, Each
Thermo Scientific Chemicals

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Catalog number: A10280.AP
500 mL, Each
Quantity

Chemical Identifiers

CAS
108-18-9
IUPAC Name
bis(propan-2-yl)amine
Molecular Formula
C6H15N
InChI Key
UAOMVDZJSHZZME-UHFFFAOYSA-N
SMILES
CC(C)NC(C)C
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms (non-U.S. specification)
Refractive Index
1.3905-1.3935 @ 20°C (non-U.S. specification)
Assay (GC)
≥99.0%

Description

Diisopropylamine is used as a precursor to prepare lithium diisopropylamide and sulfenamides, which are used in rubber vulcanization. It is also involved in the synthesis of N,N-Diisopropylethylamine (Hünig's base) obtained by alkylation with diethyl sulfate. Diisopropylammonium bromide is a bromide salt of DIPA used as an inert alternative to barium titanate. It is widely used as solvent as well as an intermediate in the the synthesis of pharmaceuticals, dyes, mineral flotation agents and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropylamine is used as a precursor to prepare lithium diisopropylamide and sulfenamides, which are used in rubber vulcanization. It is also involved in the synthesis of N,N-Diisopropylethylamine (Hünig′s base) obtained by alkylation with diethyl sulfate. Diisopropylammonium bromide is a bromide salt of DIPA used as an inert alternative to barium titanate. It is widely used as solvent as well as an intermediate in the the synthesis of pharmaceuticals, dyes, mineral flotation agents and emulsifiers.

Solubility
Miscible with water.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and plastics.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text