Thermo Scientific Chemicals

3-Fluoro-o-xylene, 99%, Thermo Scientific Chemicals

Catalog number: A10637.06
5 g, Each
Thermo Scientific Chemicals

3-Fluoro-o-xylene, 99%, Thermo Scientific Chemicals

Catalog number: A10637.06
5 g, Each
Quantity
Catalog number: A10637.06
also known as A10637-06
Price (USD)
Price: 71.00
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
443-82-3
IUPAC Name
1-fluoro-2,3-dimethylbenzene
Molecular Formula
C8H9F
InChI Key
AWLDSXJCQWTJPC-UHFFFAOYSA-N
SMILES
CC1=CC=CC(F)=C1C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.4835-1.4865 @ 20?C

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text