Thermo Scientific Chemicals

D-(+)-Xylose, 98+%

Catalog number: A10643.36
500 g, Each
Thermo Scientific Chemicals

D-(+)-Xylose, 98+%

Catalog number: A10643.36
500 g, Each
Quantity
Catalog number: A10643.36
also known as A10643-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
58-86-6
IUPAC Name
oxane-2,3,4,5-tetrol
Molecular Formula
C5H10O5
InChI Key
SRBFZHDQGSBBOR-UHFFFAOYNA-N
SMILES
OC1COC(O)C(O)C1O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
144-152°C

Description

D-(+)-Xylose is used to prepare furfural by acid catalyzed degradation reaction, which acts as a solvent as well as a precursor to synthetic polymers. It is a useful animal medicine involved in the treatment of malabsorption. It finds application in the production of sugar substitute xylitol by subjecting xylose to calyticl hydrogenation process. It is found that it undergoes an atmospheric-pressure enzyme (polyphosphate xylulokinase) driven process to get hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Xylose is used to prepare furfural by acid catalyzed degradation reaction, which acts as a solvent as well as a precursor to synthetic polymers. It is a useful animal medicine involved in the treatment of malabsorption. It finds application in the production of sugar substitute xylitol by subjecting xylose to calyticl hydrogenation process. It is found that it undergoes an atmospheric-pressure enzyme (polyphosphate xylulokinase) driven process to get hydrogen.

Solubility
Soluble in water.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text